Anti-NR5A2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106024
89354-484EA 557.47 USD
89354-484
Anti-NR5A2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to NR5A2 (nuclear receptor subfamily 5, group A, member 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NR5A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR