Anti-NOTCH1A Goat Polyclonal Antibody

Supplier: GeneTex
GTX48504
89363-658EA 656.9 USD
89363-658
Anti-NOTCH1A Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Notch1a (zebrafish)

Not yet tested. At this stage we are dependent on researchers in the field for further characterization of this product. Therefore we cannot recommend an optimal concentration and the product is investigative grade. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates?

Type: Primary
Antigen: NOTCH1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR