Anti-NOS-III(ENOS) Rabbit Polyclonal Antibody

GTX74170
89365-404EA 274.48 USD
89365-404
Anti-NOS-III(ENOS) Rabbit Polyclonal Antibody
Recognizes eNOS, a 135kD protein, in human, rat and mouse material.

Type: Primary
Antigen: NOS-III(ENOS)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR