Anti-N-MYC Mouse Monoclonal Antibody [clone: NMYC-1]

Supplier: GeneTex
GTX20057
89360-764EA 580.18 USD
89360-764
Anti-N-MYC Mouse Monoclonal Antibody [clone: NMYC-1]
Antibodies
Members of the Myc gene (c-Myc, n-Myc, and L-Myc) family code for proteins that homodimerize but do not bind well to DNA. Myc-Max heterodimers, however, bind with great affinity to the sequence CACGTG and act to initiate transcription of target genes.

Recommended Starting Dilutions:

For WB: Use at a dilution of 1:500-1:1000.

For ChIP: Use at a dilution of 1:10-1:500.

For GS: Use at a dilution of 1:1-1:100.

For ICC/IF: Use at a dilution of 1:50-1:100.

For IP: Use at a dilution of 1:10-1:100.

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: N-MYC
Clonality: Monoclonal
Clone: NMYC-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR