Anti-NMUR2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX71489
89365-270EA 590.3 USD
89365-270
Anti-NMUR2 Rabbit Polyclonal Antibody
Antibodies
Neuromedin U Receptor 2 is a Neuromedin Receptor. NMU2R may mediate the contractile activity of neuromedin U in the uterus. Neuromedin U Receptor 2 has been reported in the central nervous system and in the uterus. ESTs have been isolated from diseased brain and colon carcinoma libraries.

Type: Primary
Antigen: NMUR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR