Anti-NKIAMRE Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX70373
89364-940EA 574.44 USD
89364-940
Anti-NKIAMRE Rabbit Polyclonal Antibody
Antibodies
A novel cyclin-dependent kinase-related molecule, NKIAMRE, has been identified that is thought to be a tumor suppressor locus. The NKIAMRE gene has a cytolocalization on 5q, which is a region that is interstitially deleted in human acute leukemia and human myeloid dysplastic disorders.

Type: Primary
Antigen: NKIAMRE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR