Anti-PPP1R8 Goat Polyclonal Antibody

Supplier: GeneTex
GTX25300
89367-586EA 615.77 USD
89367-586
Anti-PPP1R8 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to NIPP1 / PPP1R8

Approx 40 kDa band observed in Human Lung and H460 cell extracts (predicted MW of 40kDa according to NP_054829). Recommended for use at 1-2µg/ml. This antibody is expected to recognise all three human isoforms of this protein.

Type: Primary
Antigen: PPP1R8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR