Anti-CHRNB2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113654
89357-672EA 557.66 USD
89357-672
Anti-CHRNB2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to AChR beta 2

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CHRNB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR