Anti-NEK7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX71879
89365-342EA 481.52 USD
89365-342
Anti-NEK7 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to Human NIMA ( Never In Mitosis Gene A ) - Related Kinase 7 ( NEK7 )

Immunohistochemistry with formalin fixed paraffin embedded tissues. Not tested for other uses. IHC-P: Use at a dilution of 1/100 - 1/200.

Type: Primary
Antigen: NEK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR