Anti-NDRG1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX41727
89367-724EA 589.25 USD
89367-724
Anti-NDRG1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to NDRG1

Approx 48kDa band observed in lysates of Human Brain, Kidney and Testes (calculated MW of 42.8kDa according to NP_006087.2). Recommended concentration: 0.1-0.3µg/ml. A minor band of unknown identity was also consistently observed at 20kDa. This band was successfully blocked by incubation with the immunising peptide. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported? The variants (NP_006087.2; NP_001128714.1) encode the same protein

Type: Primary
Antigen: NDRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR