Anti-NCF1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX20795
89360-908EA 608.44 USD
89360-908
Anti-NCF1 Goat Polyclonal Antibody
Antibodies
NCF1 is a cytosolic subunit of the multi-protein complex known as NADPH oxidase found in neutrophils.

Western Blot - Approx 47kDa band seen in U937 cell lysates. Recommended for use at 0.2-2?g/ml.

Type: Primary
Antigen: NCF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR