Anti-TBK1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX28121
89362-730EA 660.41 USD
89362-730
Anti-TBK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to NAK

For WB: Use at a dilution of 0.5 to 1 ?g/ml and an84 kDa band should be detected. Not tested in other applications. Optimal concentrations/dilutions should be determined by the researcher.

Type: Primary
Antigen: TBK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR