Anti-KAT8 Mouse Monoclonal Antibody [clone: 8C4C4]

Supplier: GeneTex
GTX83065
89353-888EA 618.49 USD
89353-888
Anti-KAT8 Mouse Monoclonal Antibody [clone: 8C4C4]
Antibodies
Mouse monoclonal antibody [8C4C4] to MYST1

Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.IHC: 1/200 - 1/1000.ICC: 1/200 - 1/1000.Not yet tested in other applications.Determining optimal working dilutions by titration test.

Type: Primary
Antigen: KAT8
Clonality: Monoclonal
Clone: 8C4C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR