Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Myosin VI Rabbit Polyclonal Antibody

Supplier: Genetex

GTX78218
89366-556EA 382.68 USD InStock
89366-556
Anti-Myosin VI Rabbit Polyclonal Antibody
Antibodies
Myosin-VI is one of the unconventional members of the myosin molecular motor superfamily. The protein has broad tissue expression and has been implicated in endocytosis at both the clathrin-coated and uncoated stages. Regulation of myosin-6 transport has been shown to involve adaptor proteins including GIPC, which contains a PDZ-domain for uncoated vesicle recruitment and Dab2 for clathrin-coated vesicle association. In hair cells of the inner ear, absence of functional myosin-VI protein leads to cell death and deafness.

Type: Primary
Antigen: Myosin VI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty