Anti-MBP Mouse Monoclonal Antibody [clone: B103.1]

GTX72695
89353-048EA 400.01 USD
89353-048
Anti-MBP Mouse Monoclonal Antibody [clone: B103.1]
Mouse Monoclonal [B103.1] to Myelin Basic Protein - (MBP)

This antibody can be used on formalin-fixed, paraffin-embedded tissue sections. Prolonged fixation in buffered formalin can destroy the epitope. The antibody may be used at a dilution of 1:25 to 1:50 with AutoProbe III (GTX28620). When using paraffin-embedded and formalin-fixed tissues, high temperature antigenic unmasking technique (see instructions below) is strongly recommended for consistent and reproducible results. This product can also be used on frozen tissue sections. The optimal conditions should be determined by the individual laboratory.

Type: Primary
Antigen: MBP
Clonality: Monoclonal
Clone: B103.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR