Anti-Mycobacterium Guinea Pig Polyclonal Antibody

Supplier: GeneTex
GTX82978
89366-766EA 376.99 USD
89366-766
Anti-Mycobacterium Guinea Pig Polyclonal Antibody
Antibodies
Mycobacterium is a genus of actinobacteria, given its own family, the Mycobacteriaceae. It includes many pathogens known to cause serious diseases in mammals, including tuberculosis and leprosy. Mycobacteria can be classified into several major groups for purpose of diagnosis and treatment: M. tuberculosis complex which can cause tuberculosis: M. tuberculosis, M. bovis, M. africanum, and M. micoti. M. leprae which causes Hansen's disease or leprosy. Nontuberculous mycobacteria (NTM) are all the other mycobacteria which can cause pulmonary disease resembling tuberculosis, lymphadenitis, skin disease, or disseminated disease.

Type: Primary
Antigen: Mycobacterium
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR