Anti-MX2 Rabbit Polyclonal Antibody

GTX102775
89349-726EA 177.66 USD
89349-726
Anti-MX2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to MX2 (myxovirus (influenza virus) resistance 2 (mouse))

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: MX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR