Anti-MVP Mouse Monoclonal Antibody [clone: 1032]

Supplier: GeneTex
GTX22376
89351-788EA 558.84 USD
89351-788
Anti-MVP Mouse Monoclonal Antibody [clone: 1032]
Antibodies
MVP is identical to Lung-Resistance Related Protein (LRP). Treatment of cells with estradiol increases the amount of MVP in nuclear extract. Antibodies to progesterone and glucocorticoid receptors are also able to co-immunoprecipitate the MVP.

For IHC-P: Use at a dilution of 1:50-1:100 in an ABC method for 30 mins at RT. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining. For IP: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MVP
Clonality: Monoclonal
Clone: 1032
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR