Anti-MUC5AC Mouse Monoclonal Antibody [clone: SPM297]

GTX18097
89351-442EA 399.48 USD
89351-442
Anti-MUC5AC Mouse Monoclonal Antibody [clone: SPM297]
Mucins are high molecular eight glycoproteins with 80% carbohydrate contents and the remaining 20% is constituted by protein core. Gastric mucin M1 antigens are detected/found in columnar mucus cells of surface gastric epithelium and in goblet cells of the fetal and precancerous colon but not in those of normal colon. Evidence from the literature suggestes that they are associated with the peptide core of mucins Resurgence of gastric mucin reactivity during colonic carcinogenesis is suggested to be due to either re-expression of the peptide core of gastric (or fetal colonic) mucins in the adult colon or due to changes in the glycosylation pattern of mucin which expose the hidden M1 antigens.

For IHC-P: Ready to use for 30 min at RT. No special pretreatment is required. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MUC5AC
Clonality: Monoclonal
Clone: SPM297
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR