Anti-MUC1 Mouse Monoclonal Antibody [clone: M2F1]

Supplier: GeneTex
GTX10118
89350-642EA 666.2 USD
89350-642
Anti-MUC1 Mouse Monoclonal Antibody [clone: M2F1]
Antibodies
Mouse monoclonal [M2F1] to MUC1

Recommended Starting Dilutions: For ELISA: Use at an assay dependent dilution. For ICC/IF: Use at an assay dependent dilution. For IHC: Use at an assay dependent dilution. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: M2F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR