Anti-MUC1 Mouse Monoclonal Antibody [clone: M10G4]

Supplier: GeneTex
GTX10115
89350-636EA 658.88 USD
89350-636
Anti-MUC1 Mouse Monoclonal Antibody [clone: M10G4]
Antibodies
Mouse monoclonal [M10G4] to MUC1

ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: M10G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR