Anti-OPRM1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX17934
89360-348EA 457.53 USD
89360-348
Anti-OPRM1 Rabbit Polyclonal Antibody
Antibodies
Most actions of exogenous opioids, such as morphine, are mediated through the μ-opioid receptor, including analgesia, tolerance and reward. In general, opioids modulate numerous central and peripheral processes including pain perception, neuroendocrine secretion and the immune response. The opioid signal is transduced from receptors through G proteins to various different effectors. Subsequent to G protein activation, several effectors are known to orchestrate the opioid signal. For example, activation of opioid receptors increases phosphatidylinositol turnover, activates K+ channels and reduces adenylyl cyclase and Ca++ channel activities.

WB: 1/1000. Predicted molecular weight: ~45 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: OPRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR