Anti-MRP2 Mouse Monoclonal Antibody [clone: M2I-4]

Supplier: GeneTex
GTX23372
89351-994EA 611.76 USD
89351-994
Anti-MRP2 Mouse Monoclonal Antibody [clone: M2I-4]
Antibodies
Mouse monoclonal [M2I - 4] to MRP2

ICC/IF:Acetone fixed cytospin preparations:Use at a 1:20-1:50 dilution.IHC:Acetone-fixed frozen sections of human tissues:Use at a 1:20 dilution.Follow by incubating with rabbit anti-mouse IgG and a monoclonal mouse APAAP complex.Alternatively,slides can be incubated with biotinylated rabbit anti-mouse IgG and streptavidin conjugated to horseradish peroxidase.

WB:Use at a 1:20-1:50 dilution and anti-mouse HRP.

Optimal dilutions / concentrations should be determined by the end user.

Type: Primary
Antigen: MRP2
Clonality: Monoclonal
Clone: M2I-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR