Anti-MRP1 Mouse Monoclonal Antibody [clone: QCRL3]

Supplier: GeneTex
GTX23370
89351-992EA 584.54 USD
89351-992
Anti-MRP1 Mouse Monoclonal Antibody [clone: QCRL3]
Antibodies
Mouse monoclonal [QCRL3] to MRP1

IHC: Use at an assay dependent dilution. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MRP1
Clonality: Monoclonal
Clone: QCRL3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR