Anti-MPP2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103908
89350-142EA 557.66 USD
89350-142
Anti-MPP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MPP2 (membrane protein, palmitoylated 2 (MAGUK p55 subfamily member 2))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MPP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR