Anti-MPO Mouse Monoclonal Antibody [clone: 2C7]

Supplier: GeneTex
GTX11730
89358-756EA 501.95 USD
89358-756
Anti-MPO Mouse Monoclonal Antibody [clone: 2C7]
Antibodies
Myeloperoxidase is a hemoprotein that is abundantly expressed in neutrophils and secreted during their activation. Native Myeloperoxidase is represented as a covalently bound tetrameric complex of two glycosylated alpha chains (MW 59 - 64 kDa) and two unglycosylated beta chains (MW 14 kDa) with total MW 150 kDa and theoretical pI 9.2. Traditionally Myeloperoxidase was considered as a main target of anti-neutrophil cytoplasm antibodies (ANCA), the serological markers for certain systemic vasculities e.g. periarteriitis nodosa, microscopic polyarteriitis and pulmonary eosinophilic granulomatosis (Churg-Strauss syndrome). Low to moderate anti-Myeloperoxidase autoantibody levels are also reported in rheumatoid arthritis. Recently it was shown that Myeloperoxidase participates in the initiation and progression of cardiovascular disease. It possesses potent proinflammatory properties and may contribute directly to tissue injury. Now Myeloperoxidase is under consideration as one of the most promising cardiac markers.

Type: Primary
Antigen: MPO
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR