Anti-MPG Goat Polyclonal Antibody

Supplier: GeneTex
GTX89783
89367-014EA 607.87 USD
89367-014
Anti-MPG Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to MPG

Approx 37kDa band observed in lysates of HEK293 and K562 (calculated MW of 32.9kDa according to NP_002425.2). Recommended concentration: 1-3µg/ml. This antibody is expected to recognize all three reported isoforms (NP_002425.2, NP_001015052.1 and NP_001015054.1)

Type: Primary
Antigen: MPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR