Anti-Mouse Goat Polyclonal Antibody

Supplier: GeneTex
GTX77264
89366-190EA 189.39 USD
89366-190
Anti-Mouse Goat Polyclonal Antibody
Antibodies
This antiserum forms multiple arcs against normal mouse serum by immunoelectrophoresis.. Goats were immunized with pooled normal mouse serum in Freund's adjuvant

Type: Secondary
Antigen: Mouse Whole Serum
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR