Anti-MOP3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30121
89352-650EA 617.24 USD
89352-650
Anti-MOP3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to MOP3 / BMAL1

WB: Use at a dilution of 1/100 - 1/200. May be used in Western analysis where it recognizes a band at ~73 kDa representing MOP3. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MOP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR