Anti-MMP9 Mouse Monoclonal Antibody [clone: 6-6B]

GTX11225
89350-812EA 377.06 USD
89350-812
Anti-MMP9 Mouse Monoclonal Antibody [clone: 6-6B]
Mouse monoclonal [6-6B] to MMP9

For IF: Use at an assay dependent dilution. For IP: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR