Anti-MMP23 Goat Polyclonal Antibody

Supplier: GeneTex
GTX73121
89353-140EA 376.85 USD
89353-140
Anti-MMP23 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to MMP - 23

For IHC-P: Use at a dilution of 1:1,000. Not tested in other applications. Optimal dilutions/concentrtaions should be determined by the researcher.

Type: Primary
Antigen: MMP23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR