Anti-MLL4 Goat Polyclonal Antibody

Supplier: GeneTex
GTX89763
89367-012EA 607.87 USD
89367-012
Anti-MLL4 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to MLL4

Preliminary experiments gave no signal but low background in Human Heart lysates at up to 1µg/ml. However this used our routine western blotting protocol which we would not expect to reliably detect proteins as large as the predicted size of approx. 312KDa according to NP_055542. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Immunohistochemistry: In paraffin embedded Human Testis shows cytoplasm staining of spermatids. Recommended concentration, 5-10µg/ml.

Type: Primary
Antigen: MLL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR