Anti-MKK3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX25428
89361-636EA 631.81 USD
89361-636
Anti-MKK3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to MKK3

For IP and WB: Use at a dilution of 1:1,500 - 1:2,500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MKK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR