Anti-MIP2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX74085
89367-752EA 617.12 USD
89367-752
Anti-MIP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CXCL2

Recommended Starting Dilutions: ELISA: Max Dilution 0.5 ug/ml Functional Assays: 0.50 ug/ml - 0.90 ug/ml Western Blotting: 0.1 ug/ml - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by

Type: Primary
Antigen: MIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR