Anti-MK Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12739
89359-508EA 611 USD
89359-508
Anti-MK Rabbit Polyclonal Antibody
Antibodies
Midkine exhibits neurite outgrowth-promoting activity and may play a role in nervous system development and/or maintenance. Its expression is predominant only for a short period from approximately one-half to two-thirds of the way through gestation; before and after that, it is barely detectable. Midkine was first found in differentiating mouse teratocarcinoma cells. It has neurotrophic activities and is mitogenic to certain, but not to all, fibroblast cell lines.

Type: Primary
Antigen: MK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR