Anti-MHC CLASS I RT1A Mouse Monoclonal Antibody

Supplier: GeneTex
GTX75168
89365-710EA 595.9 USD
89365-710
Anti-MHC CLASS I RT1A Mouse Monoclonal Antibody
Antibodies
gene complex that may play a role in antigen presentation [RGD]

Type: Primary
Antigen: MHC CLASS I RT1A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR