Anti-GRM3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10309
89349-848EA 585.53 USD
89349-848
Anti-GRM3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to metabotropic Glutamate Receptor 3

WB: Use at a dilution of 1/1000. Predicted molecular weight: 105.2 kDa. GTX10309 has only been shown to work by Western blot in systems using highly expressing transfected cells using standard protocols. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Grm3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR