Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Gosr2 Mouse Monoclonal Antibody [clone: 4HAD6 ]

Supplier: Genetex
GTX13511
Membrin is a 27 kDa integral membrane protein on the surface of the Golgi apparatus that serves as a tSNARE in ER to Golgi transport. GS27 is a member of a protein complex that consists of cis-Golgi vesicle receptor syntaxin 5 and GS28, vSNARE rbet1 and rsec22, and rsly1. Immunoprecipitation analysis has shown that there are two subcomplexes within this complex: one contains syntaxin 5 and GOS28 and the other one contains syntaxin 5, GS27/membrin, rsec22 and rbet1. These complexes are postulated to direct the fusion of ER-derived vesicles with Vesicular tubular cluster (VTC), and the fusion of VTCs to form cis-Golgi compartment.

ICC: Use at a dilution of 1/500. IP: Use at a concentration of 2 - 4 μg/ml. WB: Use at a concentration of 2 μg/ml. This concentration detects membrin in 20μg of HeLa heat shocked cell lysate by ECL analysis. Predicted molecular weight: 27 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Gosr2
Clonality: Monoclonal
Clone: 4HAD6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Dog, Pig, Rat, Hamster, Rabbit




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 μg GTX13511 89359-710 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol GOSR2
Antigen Name Golgi SNAP Receptor Complex member 2
Reactivity Dog
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 4HAD6
Host Mouse
Isotype IgG1
Western Blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Antigen Synonyms GS27, EPM6, Bos1, Membrin
Gene ID 64154
Concentration 1 mg/ml
Immunogen Recombinant rat membrin with a His6 tag
Purification Protein G purified
Storage Buffer Preservative: NoneConstituents: 50% Glycerol, 0.1mM PMSF, PBS. pH 7.2
Storage Temperature Keep as concentrated solution. Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
Tested Applications ICC/IF, IP, WB

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty