Anti-MECP2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX82658
89366-736EA 460.15 USD
89366-736
Anti-MECP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MeCP2 (phospho Ser80)

  • Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MECP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR