Anti-MDM2 Mouse Monoclonal Antibody [clone: HDM2-323]

Supplier: GeneTex
GTX10567
89354-334EA 607.14 USD
89354-334
Anti-MDM2 Mouse Monoclonal Antibody [clone: HDM2-323]
Antibodies
Mouse monoclonal [HDM2 - 323] to MDM2

ELISA: Use at an assay dependent dilution. WB: Use at a concentration of 1 - 2 ?g/ml. Detects a band of approximately 90 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MDM2
Clonality: Monoclonal
Clone: HDM2-323
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR