Anti-MDC Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX27362
89362-568EA 660.25 USD
89362-568
Anti-MDC Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to MDC

WB: Use at a dilution of 1/2000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR