Anti-MCT1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30233
89362-900EA 600.11 USD
89362-900
Anti-MCT1 Rabbit Polyclonal Antibody
Antibodies
MCT1 is a novel gene that was identified from T-cell leukemia/lymphoma cell lines with limited homology to a region of Cyclin H. MCT1 is novel candidate oncogene that results in a shortened G1 phase and increased rate of proliferation.

Type: Primary
Antigen: MCT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR