Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MCSF Mouse Monoclonal Antibody [clone: 692]

Supplier: Genetex

GTX18640
89360-486EA 358.9 USD InStock
89360-486
Anti-MCSF Mouse Monoclonal Antibody [clone: 692]
Antibodies
Four distinct colony-stimulating factors (CSFs) that promote survival, proliferation and differentiation of bone marrow precursor cells have been well characterized: granulocyte macrophage CSF (GMCSF), granulocyte CSF (GCSF), macrophage CSF (MCSF), and Interleukin-3 (IL-3, Multi CSF). Both GMCSF and IL-3 are multipotential growth factors, stimulating proliferation of progenitor cells from more than one hematopoietic lineage. In contrast, GCSF and MCSF are lineage restricted hematopoietic growth factors, stimulating final mitotic divisions and the terminal cellular maturation of the partially differentiated hematopoietic progenitors. Macrophage CSF, also known as CSF1, is produced by monocytes, fibroblasts and endothelial cells. It stimulates the formation of macrophage colonies, enhances antibody-dependent, cell-mediated cytotoxicity by monocytes and macrophages, and inhibits bone resorption by osetoclasts. Natural human MCSF is a dimeric glycoprotein of 70-90 kD molecular weight, existing in multiple glycosylation forms. It binds to a 165 kD glycoprotein of the receptor tyrosine kinase subclass III, a family that includes the receptors for platelet derived growth factor (PDGF) and stem cell factor (SCF).

ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MCSF
Clonality: Monoclonal
Clone: 692
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty