Anti-MCM2 Mouse Monoclonal Antibody [clone: CRCT2.1]

Supplier: GeneTex
GTX75459
89367-880EA 615.77 USD
89367-880
Anti-MCM2 Mouse Monoclonal Antibody [clone: CRCT2.1]
Antibodies
Mouse monoclonal antibody [CRCT2.1] to MCM2

Recommended Starting Dilutions: Immunohistology - Frozen: 1/500 - 1/1000 Immunohistology - Paraffin: Max Dilution 1/200 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: MCM2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR