Anti-MAX Goat Polyclonal Antibody

Supplier: GeneTex
GTX89997
89367-018EA 607.87 USD
89367-018
Anti-MAX Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to MAX

Approx 23kDa band seen in Jurkat Lysates. Recommended for use at 0.5-2 µg/ml. This antibody is expected to recognize both reported isoforms (NP_002373.3; NP_660087.1).

Type: Primary
Antigen: MAX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR