Anti-MAST1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107508
89354-982EA 582.76 USD
89354-982
Anti-MAST1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MAST1 (microtubule associated serine/threonine kinase 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MAST1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR