Anti-GPCR GPR80/GPR99 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX71875
89365-338EA 481.52 USD
89365-338
Anti-GPCR GPR80/GPR99 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to Human G Protein-Coupled Receptor
GPR80/GPR99/P2Y15

For IHC-P: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: OXGR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR