Anti-LYN Mouse Monoclonal Antibody [clone: LYN-01]

Supplier: GeneTex

GTX21890
89361-104EA 427.48 USD
89361-104
Anti-LYN Mouse Monoclonal Antibody [clone: LYN-01]
Antibodies
Lyn is - member of the src family of non-receptor protein tyrosine kinases that is predominantly expressed in haematopoietic tissues. Like all members of the src family, lyn is thought to participate in signal transduction from cell surface receptors that lack intrinsic tyrosine kinase activity. It is associated with - number of cell surface receptors including the - cell antigen receptor and immunoglobulin - receptor (FceRI).

ICC: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 58 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LYN
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR