Anti-LXR beta Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX113571
89357-624EA 427.1 USD
89357-624
Anti-LXR beta Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to LXR beta (nuclear receptor subfamily 1, group H, member 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LXR beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR