Anti-LSM1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX47320
89363-624EA 612.19 USD
89363-624
Anti-LSM1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to LSM1 (Middle)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.<p>Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: LSM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR